Få mere at vide
Methyl-d3 Paraben, TRC
Artikelnummer. 30482073
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30482073 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482073 Leverandør Toronto Research Chemicals Leverandørnr. TRCM325662100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl-D3 4-Hydroxybenzoate
CAS 1216543-26-8
Molekylær formel C8 D3 H5 O3
Mængde 100 mg
Synonym Methyl-d3 Paraben, Trideuteriomethyl 4-hydroxybenzoate, Methyl-d3 Paraben, Methyl 4-hydroxybenzoate-d3, Methyl-D3 4-hydroxybenzoate, Methyl parahydroxybenzoate-d3, 4-Hydroxybenzoic acid methyl-d3 ester, Methylparaben-(methyl-d3)
InChI formel InChI=1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3/i1D3
SMIL [2H]C([2H])([2H])OC(=O)c1ccc(O)cc1
IUPAC navn trideuteriomethyl 4-hydroxybenzoate
Molekylvægt (g/mol) 155.166
Formel vægt 155.066