Få mere at vide
Methyl-d3 Methacrylate, CDN
Artikelnummer. 30317357
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317357 0.5 g 0.5g
30310404 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317357 Leverandør CDN Leverandørnr. CDND24730.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methacrylic Acid Methyl Ester-D3 (methoxy-D3)
CAS 35777-12-9
Molekylær formel C5 D3 H5 O2
Mængde 0.5 g
Synonym 2-Propenoic acid, 2-methyl-, methyl-d3 ester (9 CI), Methyl-d3 methacrylate, Methacrylic acid methyl-d3 ester, 2-Methyl-2-propenoic acid methyl-d3 ester, Methacrylic acid methyl ester-D3 (methoxy-D3)
InChI formel InChI=1 S/C5H8O2/c1-4(2)5(6)7-3/h1H2,2-3H3/i3D3
SMIL [2 H]C([2 H])([2 H])OC(=O)C(=C)C
IUPAC navn trideuteriomethyl 2-methylprop-2-enoate
Molekylvægt (g/mol) 103.1343
Formel vægt 103.0713 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity