Få mere at vide
Methyl-d3-malonic Acid, CDN
Artikelnummer. 30318784
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318784 0.25 g -
30316237 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318784 Leverandør CDN Leverandørnr. CDND28100.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl-D3-malonic acid
CAS 42522-59-8
Molekylær formel C4 2H3 H3 O4
Mængde 0.25 g
InChI formel InChI=1 S/C4H6O4/c1-2(3(5)6)4(7)8/h2H,1H3,(H,5,6)(H,7,8)/i1D3
SMIL [2 H]C([2 H])([2 H])C(C(=O)O)C(=O)O
IUPAC navn 2-(trideuteriomethyl)propanedioic acid
Molekylvægt (g/mol) 121.11
Formel vægt 121.0454 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity