Få mere at vide
Methyl-d3 Formate, CDN
Artikelnummer. 30326595
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326595 1 g
30317260 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326595 Leverandør CDN Leverandørnr. CDND02481G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Formate-D3 (methyl-D3)
CAS 23731-39-7
Molekylær formel C2 D3 H O2
Mængde 1 g
Synonym Methan-d3-ol, formate (9 CI), Formic acid, methyl-d3 ester (6 CI, 7 CI, 8 CI), Methyl-d3 formate, Formic acid methyl-d3 ester, Methyl formate-D3 (methyl-D3)
InChI formel InChI=1 S/C2H4O2/c1-4-2-3/h2H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])OC=O
IUPAC navn trideuteriomethyl formate
Molekylvægt (g/mol) 63.0705
Formel vægt 63.04 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity