Få mere at vide
Methyl-d3- 2,3,4-Tri-O-acetyl-b-D-glucuronic Acid Methyl Ester, TRC
Artikelnummer. 30470539
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30470539 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470539 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33082250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl-d3- 2,3,4-Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester
CAS 72661-75-7
Molekylær formel C14H17D3O10
Mængde 50 mg
Synonym Methyl-d3 Triacetate-beta-D-glucopyranosiduronic Acid Methyl Ester
InChI formel InChI=1S/C14H20O10/c1-6(15)21-9-10(22-7(2)16)12(23-8(3)17)14(20-5)24-11(9)13(18)19-4/h9-12,14H,1-5H3/t9-,10-,11-,12+,14+/m0/s1/i5D3
SMIL [2H]C([2H])([2H])O[C@@H]1O[C@@H]([C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C)C(=O)OC
IUPAC navn methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-(trideuteriomethoxy)oxane-2-carboxylate
Molekylvægt (g/mol) 351.32
Formel vægt 351.1245