Få mere at vide
Methyl D-Pantothenate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl Pantothenate |
| CAS | 50692-78-9 |
| Molekylær formel | C10 H19 N O5 |
| Mængde | 500 mg |
| Synonym | N-[(2R)-2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl]-β-alanine methyl ester, Pantothenic acid, methyl ester (6CI), β-Alanine, N-(2,4-dihydroxy-3,3-dimethyl-1-oxobutyl)-, methyl ester, (R)-, D-Pantothenic acid methyl ester, Methyl D-pantothenate, Methyl pantothenate |
| InChI formel | InChI=1S/C10H19NO5/c1-10(2,6-12)8(14)9(15)11-5-4-7(13)16-3/h8,12,14H,4-6H2,1-3H3,(H,11,15)/t8-/m0/s1 |
| SMIL | COC(=O)CCNC(=O)[C@H](O)C(C)(C)CO |
| IUPAC navn | methyl 3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate |
| Molekylvægt (g/mol) | 233.26 |
| Formel vægt | 233.1263 |