Få mere at vide
Methyl D-(-)-4-Hydroxy-phenylglycinate, TRC
Artikelnummer. 30480457
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480457 100 mg
30470137 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30480457 Leverandør Toronto Research Chemicals Leverandørnr. TRCM338538100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 37763-23-8
Molekylær formel C9 H11 N O3
Molekylvægt (g/mol) 181.19
Synonym Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, (αR)-, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, (R)-, (R)-Methyl (4-hydroxyphenyl)glycinate, (R)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate, D-(4-Hydroxyphenyl)glycine methyl ester, D-p-Hydroxyphenylglycine methyl ester, Methyl (R)-2-amino-2-(4-hydroxyphenyl)acetate, Methyl D-(-)-α-amino-α-(4-hydroxyphenyl)acetate, HPGMe, HPGMe, HPG, HPGMe Test Mix, Amoxicillin trihydrate (Enzymatic) related
IUPAC navn methyl (2R)-2-amino-2-(4-hydroxyphenyl)acetate
SMIL COC(=O)[C@H](N)c1ccc(O)cc1
Tekniske data
Kemisk navn eller materiale D-p-Hydroxyphenylglycine Methyl Ester
Mængde 100 mg
InChI formel InChI=1S/C9H11NO3/c1-13-9(12)8(10)6-2-4-7(11)5-3-6/h2-5,8,11H,10H2,1H3/t8-/m1/s1
Formel vægt 181.0739