missing translation for 'onlineSavingsMsg'
Learn More

Methyl chlorodifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-7024268
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1514-87-0
C3H3ClF2O2
144.502
MFCD00000775
AWUPLMYXZJKHEG-UHFFFAOYSA-N
methyl chlorodifluoroacetate, chlorodifluoroacetic acid methyl ester, acetic acid, chlorodifluoro-, methyl ester, methyl difluorochloroacetic acid, methyl chloro difluoro acetate, acetic acid, 2-chloro-2,2-difluoro-, methyl ester, methyl-2-chloro-2,2-difluoroacetate, chloro-difluoro-acetic acid methyl ester, clcf2coome, clcf2co2me
73935
methyl 2-chloro-2,2-difluoroacetate
COC(=O)C(F)(F)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1514-87-0
C3H3ClF2O2
144.502
MFCD00000775
AWUPLMYXZJKHEG-UHFFFAOYSA-N
methyl chlorodifluoroacetate, chlorodifluoroacetic acid methyl ester, acetic acid, chlorodifluoro-, methyl ester, methyl difluorochloroacetic acid, methyl chloro difluoro acetate, acetic acid, 2-chloro-2,2-difluoro-, methyl ester, methyl-2-chloro-2,2-difluoroacetate, chloro-difluoro-acetic acid methyl ester, clcf2coome, clcf2co2me
73935
methyl 2-chloro-2,2-difluoroacetate
COC(=O)C(F)(F)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.371
Boiling Point 79°C to 81°C
Flash Point 19°C (66°F)
Refractive Index 1.35
Quantity 10 g
UN Number UN2924
Beilstein 1756072
Sensitivity Moisture sensitive
Formula Weight 144.5
Percent Purity 98%
Chemical Name or Material Methyl chlorodifluoroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'