missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl chlorodifluoroacetate, 98%

Brand:  Alfa Aesar™ A18623.18

Additional Details : CAS Number : 1514-87-0 Weight : 0.05000kg Transport : UN number : 2924 Chem class : 3 Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11498557

  • 739.00 DKK / 50g

Estimated Shipment
17-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1514-87-0
144.502
AWUPLMYXZJKHEG-UHFFFAOYSA-N
73935
COC(=O)C(F)(F)Cl
C3H3ClF2O2
MFCD00000775
methyl chlorodifluoroacetate, chlorodifluoroacetic acid methyl ester, acetic acid, chlorodifluoro-, methyl ester, methyl difluorochloroacetic acid, methyl chloro difluoro acetate, acetic acid, 2-chloro-2,2-difluoro-, methyl ester, methyl-2-chloro-2,2-difluoroacetate, chloro-difluoro-acetic acid methyl ester, clcf2coome, clcf2co2me
methyl 2-chloro-2,2-difluoroacetate

Specifications

Methyl chlorodifluoroacetate
19°C (66°F)
Moisture sensitive
50g
C3H3ClF2O2
UN2924
methyl chlorodifluoroacetate, chlorodifluoroacetic acid methyl ester, acetic acid, chlorodifluoro-, methyl ester, methyl difluorochloroacetic acid, methyl chloro difluoro acetate, acetic acid, 2-chloro-2,2-difluoro-, methyl ester, methyl-2-chloro-2,2-difluoroacetate, chloro-difluoro-acetic acid methyl ester, clcf2coome, clcf2co2me
COC(=O)C(F)(F)Cl
144.502
144.5
1.371
1.35
79°C to 81°C
1514-87-0
MFCD00000775
1756072
AWUPLMYXZJKHEG-UHFFFAOYSA-N
methyl 2-chloro-2,2-difluoroacetate
73935
98%
Videos
SDS Documents