Få mere at vide
Methyl Bexarotenate, TRC
Artikelnummer. 30480440
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480440 100 mg
30476969 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30480440 Leverandør Toronto Research Chemicals Leverandørnr. TRCM289820100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 153559-48-9
Molekylær formel C25H30O2
Molekylvægt (g/mol) 362.5
Synonym Bexarotene Impurity Z4F
IUPAC navn methyl 4-[1-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethenyl]benzoate
SMIL COC(=O)c1ccc(cc1)C(=C)c2cc3c(cc2C)C(C)(C)CCC3(C)C
Tekniske data
Kemisk navn eller materiale Methyl Bexarotenate
Mængde 100 mg
InChI formel InChI=1S/C25H30O2/c1-16-14-21-22(25(5,6)13-12-24(21,3)4)15-20(16)17(2)18-8-10-19(11-9-18)23(26)27-7/h8-11,14-15H,2,12-13H2,1,3-7H3
Formel vægt 362.2246