Få mere at vide
Methyl Benzoate-2,3,4,5,6-d5, CDN
Artikelnummer. 30313973
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313973 1 g -
30322165 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313973 Leverandør CDN Leverandørnr. CDND61551G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Benzoate-2,3,4,5,6-D5
CAS 68661-19-8
Molekylær formel C8 D5 H3 O2
Mængde 1 g
Synonym Benzoic-d5 acid, methyl ester (7 CI, 9 CI, ACI), Methyl benzoate-d5 (ACI), Methyl Benzoate-2,3,4,5,6-d5, Benzoic-d5 acid methyl ester, Methyl Benzoate-2,3,4,5,6-D5, Methyl benzoate-d5 (phenyl-d5)
InChI formel InChI=1 S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6 H,1H3/i2D,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OC)c([2 H])c1[2 H]
IUPAC navn methyl 2,3,4,5,6-pentadeuteriobenzoate
Molekylvægt (g/mol) 141.1787
Formel vægt 141.0838 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity