missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl benzilate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
It is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
Specifications
Melting Point | 73°C to 76°C |
Boiling Point | 187°C (13 mmHg) |
Quantity | 50 g |
Beilstein | 2121492 |
Merck Index | 14,1080 |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 242.28 |
Percent Purity | 98% |
Chemical Name or Material | Methyl benzilate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'