Få mere at vide
Methyl Atrarate, TRC
Artikelnummer. 30472466
Skift visning
Mængde:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472466 25 g
30479285 5 g
30477411 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30472466 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28834025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4707-47-5
Molekylær formel C10 H12 O4
Molekylvægt (g/mol) 196.1999
Synonym Atraric acid (6CI), β-Resorcylic acid, 3,6-dimethyl-, methyl ester (7CI,8CI), 2,4-Dihydroxy-3,6-dimethyl-benzoic acid methyl ester, Everniate, Evernyl, LRG 201, Methyl 2,4-dihydroxy-3,6-dimethylbenzoate, Methyl 3,6-dimethyl-2,4-dihydroxybenzoate, Methyl 3,6-dimethyl-β-resorcylate, Methyl atrarate, Methyl β-orcinolcarboxylate, Methyl β-orsellinate, Moss Synth, Mousse cristal, Oakmoss synthetic, Veramoss, 2,4-Dihydroxy-3,6-dimethylbenzoic Acid Methyl Ester, Atraric acid-methyl ester, Methyl atratate
IUPAC navn methyl 2,4-dihydroxy-3,6-dimethylbenzoate
SMIL COC(=O)c1c(C)cc(O)c(C)c1O
Tekniske data
Kemisk navn eller materiale 2,4-Dihydroxy-3,6-dimethylbenzoic Acid Methyl Ester
Mængde 25 g
InChI formel InChI=1S/C10H12O4/c1-5-4-7(11)6(2)9(12)8(5)10(13)14-3/h4,11-12H,1-3H3
Formel vægt 196.0736