Få mere at vide
Methyl Arsonic Acid Disodium Salt (~40%, Contain Sodium Sulfate), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Disodium Methylarsonate |
| CAS | 144-21-8 |
| Molekylær formel | C H3 As O3 . 2 Na |
| Mængde | 1 g |
| Synonym | Arsonic acid, methyl-, disodium salt (9CI), Methanearsonic acid, disodium salt (8CI), Ansar 184, Ansar 8100, Arifen, Arrhenal, Arsenil, Arsinyl, Arsynal, Cacodyl New, Cralo-E-rad, DMA, DMA 100, DSMA, Diarsen, Disodium methanearsonate, Disodium methylarsenate(2-), Disodium methylarsonate, Disomear, Methar, Metharsan, Metharsinat, Neo-Asycodile, Sodar, Somar, Stenosine, Tonarsan, Tonarsin, Methyl Arsonic Acid Disodium Salt |
| InChI formel | InChI=1S/CH5AsO3.2Na/c1-2(3,4)5;;/h1H3,(H2,3,4,5);;/q;2*+1/p-2 |
| SMIL | [Na+].[Na+].C[As](=O)([O-])[O-] |
| IUPAC navn | disodium;methyl-dioxido-oxo-λ^{5}-arsane |
| Molekylvægt (g/mol) | 183.93 |
| Formel vægt | 183.9094 |