Få mere at vide
Methyl Alpha-L-Daunosamine Hydrochloride (Alpha:Beta approx. 85:15), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl alpha-L-Daunosamine Hydrochloride |
| CAS | 32385-06-1 |
| Molekylær formel | C7 H15 N O3 . Cl H |
| Mængde | 10 mg |
| Synonym | L-lyxo-Hexopyranoside, methyl 3-amino-2,3,6-trideoxy-, hydrochloride, α- (8CI), α-L-lyxo-Hexopyranoside, methyl 3-amino-2,3,6-trideoxy-, hydrochloride (9CI), α-Methyl daunosaminide hydrochloride, α-L-lyxo-Hexopyranoside, methyl 3-amino-2,3,6-trideoxy-, hydrochloride (1:1) |
| InChI formel | InChI=1S/C7H15NO3.ClH/c1-4-7(9)5(8)3-6(10-2)11-4;/h4-7,9H,3,8H2,1-2H3;1H/t4-,5-,6+,7+;/m0./s1 |
| SMIL | Cl.CO[C@H]1C[C@H](N)[C@H](O)[C@H](C)O1 |
| IUPAC navn | (2S,3S,4S,6R)-4-amino-6-methoxy-2-methyloxan-3-ol;hydrochloride |
| Molekylvægt (g/mol) | 197.66 |
| Formel vægt | 197.0819 |