Få mere at vide
Methyl Alpha-L-Arabinopyranoside, TRC
Artikelnummer. 30474725
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474725 5 mg
30464909 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30474725 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2882555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3945-28-6
Molekylær formel C6H12O5
Molekylvægt (g/mol) 164.16
Synonym alpha-L-Arabinopyranoside Methyl Ether
IUPAC navn (2R,3R,4S,5S)-2-methoxyoxane-3,4,5-triol
SMIL CO[C@@H]1OC[C@H](O)[C@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Methyl alpha-L-Arabinopyranoside
Mængde 5 mg
InChI formel InChI=1S/C6H12O5/c1-10-6-5(9)4(8)3(7)2-11-6/h3-9H,2H2,1H3/t3-,4-,5+,6+/m0/s1
Formel vægt 164.0685