missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl adamantane-1-carboxylate, 97%, BLD Pharm™

Artikelnummer. 31432909
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31432909 100 g
31427715 10 g
31432066 1 g
31420349 25 g
31430748 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31432909 Supplier BLD Pharm Supplier No. BD112684100g
 Begrænset vare
Forventet forsendelse: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale Methyl adamantane-1-carboxylate
CAS 711-01-3
Molekylær formel C12H18O2
MDL nummer MFCD01838519
Mængde 100 g
SMIL O=C(C12CC3CC(C2)CC(C3)C1)OC
Molekylvægt (g/mol) 194.27
Procent renhed 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.