Få mere at vide
Methyl Acetylsalicylate, Mikromol™
Artikelnummer. 30052020
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052020 25 mg
30050263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30052020 Leverandør Mikromol Leverandørnr. MM0133.120025
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl Acetylsalicylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Acetylsalicylate
CAS 580-02-9
Analyt- eller komponentnavne Methyl Acetylsalicylate
Forsendelsestilstand Room Temperature
Molekylær formel C10 H10 O4
Certificeringer/compliance ISO 17025
Synonym Methyl Acetylsalicylate, 2-(Acetyloxy)-benzoic acid methyl ester, Acetylsalicylic acid, methyl ester (6CI), Salicylic acid acetate, methyl ester (8CI), Salicylic acid, methyl ester, acetate (7CI), Aspirin methyl ester, Methyl 2-acetoxybenzoate, Methyl O-acetylsalicylate, Methyl acetylsalicylate, Methyl aspirin, Methyl o-acetoxybenzoate, Methyl rhodin, Methylrhodine, NSC 403847
InChI formel InChI=1S/C10H10O4/c1-7(11)14-9-6-4-3-5-8(9)10(12)13-2/h3-6H,1-2H3
SMIL COC(=O)c1ccccc1OC(=O)C
IUPAC navn methyl 2-acetyloxybenzoate
Molekylvægt (g/mol) 194.18
Mængde 25 mg
Formel vægt 194.0579 g/mol