Få mere at vide
Methyl Acetylricinoleate (>80%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ricinoleic Acid-Methyl Ester Acetate |
| CAS | 140-03-4 |
| Molekylær formel | C21 H38 O4 |
| Mængde | 1 mL |
| Synonym | 9-Octadecenoic acid, 12-(acetyloxy)-, methyl ester, [R-(Z)]-, Ricinoleic acid, methyl ester, acetate (6CI,7CI,8CI), Flexricin P 4, MAR-N, Methyl O-acetylricinoleate, Methyl acetyl ricinoleate, Methyl cis-(R)-12-acetoxyoctadec-9-enoate, Methyl ricinoleate |
| InChI formel | InChI=1S/C21H38O4/c1-4-5-6-13-16-20(25-19(2)22)17-14-11-9-7-8-10-12-15-18-21(23)24-3/h11,14,20H,4-10,12-13,15-18H2,1-3H3/b14-11-/t20-/m1/s1 |
| SMIL | CCCCCC[C@H](C\C=C/CCCCCCCC(=O)OC)OC(=O)C |
| IUPAC navn | methyl (Z,12R)-12-acetyloxyoctadec-9-enoate |
| Molekylvægt (g/mol) | 354.524 |
| Formel vægt | 354.277 |