Få mere at vide
Methyl Acetate-d3, CDN
Artikelnummer. 30326711
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30326711 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326711 Leverandør CDN Leverandørnr. CDND00661G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Acetate-D3 (acetyl-D3)
CAS 21300-54-9
Molekylær formel C3 D3 H3 O2
Mængde 1 g
Synonym Acetic-d3 acid, methyl ester (6 CI, 8 CI, 9 CI), Methyl acetate-d3, Methyl Acetate-D3 (acetyl-D3), Acetic-d3 acid methyl ester, Acetic acid methyl ester-d3
InChI formel InChI=1 S/C3H6O2/c1-3(4)5-2/h1-2H3/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)OC
IUPAC navn methyl 2,2,2-trideuterioacetate
Molekylvægt (g/mol) 77.097
Formel vægt 77.0556 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity