Få mere at vide
Methyl 6-Methoxy-2-naphthylacetate-d6, TRC
Artikelnummer. 30470461
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30470461 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470461 Leverandør Toronto Research Chemicals Leverandørnr. TRCM320487100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 6-Methoxy-2-naphthylacetate-d6
CAS 1246815-39-3
Molekylær formel C14H8D6O3
Mængde 100 mg
Synonym 6-Methoxy-2-naphthaleneacetic Acid Methyl Ester-d6
InChI formel InChI=1S/C14H14O3/c1-16-13-6-5-11-7-10(8-14(15)17-2)3-4-12(11)9-13/h3-7,9H,8H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])OC(=O)Cc1ccc2cc(OC([2H])([2H])[2H])ccc2c1
IUPAC navn trideuteriomethyl 2-[6-(trideuteriomethoxy)naphthalen-2-yl]acetate
Molekylvægt (g/mol) 236.3
Formel vægt 236.132