Learn More
Methyl 6-bromo-1-methylindole-2-carboxylate, 97%, Thermo Scientific™
Catalog Number p-7055148
Click to view available options
Quantity:
250 mg
1 g
5 g
680569-18-0
C11H10BrNO2
268.11
MFCD07364734
RCDWEKOOOHATJC-UHFFFAOYSA-N
methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
4777911
methyl 6-bromo-1-methylindole-2-carboxylate
CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC
680569-18-0
C11H10BrNO2
268.11
MFCD07364734
RCDWEKOOOHATJC-UHFFFAOYSA-N
methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
4777911
methyl 6-bromo-1-methylindole-2-carboxylate
CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
Formula Weight 268.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 6-bromo-1-methylindole-2-carboxylate