missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 6-Aminopyridine-3-carboxylate, TRC

Artikelnummer. 30476203
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476203 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30476203 Leverandør Toronto Research Chemicals Leverandørnr. TRCM286855250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 6-Aminopyridine-3-carboxylate
CAS 36052-24-1
Molekylær formel C7 H8 N2 O2
Mængde 250 mg
Synonym 3-Pyridinecarboxylic acid','6-amino-','methyl ester, Nicotinic acid','6-amino-','methyl ester (6CI,8CI), 2-Amino-5-(methoxycarbonyl)pyridine, 5-Methoxycarbonyl-2-Aminopyridine, 6-Aminonicotinic acid methyl ester, 6-Aminopyridine-3-carboxylic acid methyl ester, Methyl 2-aminopyridine-5-carboxylate, Methyl 6-aminonicotinate, Methyl 6-aminopyridine-3-carboxylate
InChI formel InChI=1S/C7H8N2O2/c1-11-7(10)5-2-3-6(8)9-4-5/h2-4H,1H3,(H2,8,9)
SMIL COC(=O)c1ccc(N)nc1
IUPAC navn methyl 6-aminopyridine-3-carboxylate
Molekylvægt (g/mol) 152.15
Formel vægt 152.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.