Få mere at vide
Methyl 5-Oxopyrrolidine-3-carboxylate, TRC
Artikelnummer. 30474702
Skift visning
Mængde:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474702 10 g
30469135 1 g
30465019 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30474702 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32469010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35309-35-4
Molekylær formel C6H9NO3
Molekylvægt (g/mol) 143.14
Synonym 2-Oxo-4-pyrrolidinecarboxylic Acid Methyl Ester, 5-Oxo-3-pyrrolidinecarboxylic Acid Methyl Ester
IUPAC navn methyl 5-oxopyrrolidine-3-carboxylate
SMIL O=C1NCC(C(OC)=O)C1
Tekniske data
Kemisk navn eller materiale Methyl 5-Oxopyrrolidine-3-carboxylate
Mængde 10 g
InChI formel InChI=1S/C6H9NO3/c1-10-6(9)4-2-5(8)7-3-4/h4H,2-3H2,1H3,(H,7,8)
Formel vægt 143.06