Learn More
Methyl 5-methylnicotinate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 29681-45-6 |
| Molecular Formula | C8H9NO2 |
| MDL Number | MFCD01075739 |
| Quantity | 100 g |
| SMILES | C1=C(C=NC=C1C)C(OC)=O |
| Molecular Weight (g/mol) | 151.16 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl 5-methylnicotinate |