Få mere at vide
Methyl 5-Hydroxy-2-benzimidazolecarbamate, TRC
Artikelnummer. 30011246
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011246 100 mg
30004454 10 mg
30005683 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011246 Leverandør Toronto Research Chemicals Leverandørnr. TRCM312585100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22769-68-2
Molekylær formel C9H9N3O3
Molekylvægt (g/mol) 207.19
Synonym 5-Hydroxy-2-methoxycarbonylaminobenzimidazole, 5-HydroxyMBC, 5-Hydroxycarbendazim, Methyl 5-hydroxybenzimidazol-2-ylcarbamate;
SMIL O=C(CC(OCC)=O)CCCOCC1=CC=CC=C1
Tekniske data
Kemisk navn eller materiale Methyl 5-Hydroxy-2-benzimidazolecarbamate
Renhedsgrad noter HPLC
Mængde 100 mg
Procent renhed >95%