missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 5-Ethyl-2-pyridine-carboxylate, TRC

Artikelnummer. 30472712
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472712 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472712 Supplier Toronto Research Chemicals Supplier No. TRCM304950250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 5-Ethyl-2-pyridine-carboxylate
CAS 13509-14-3
Molekylær formel C9 H11 N O2
Mængde 250 mg
Synonym 2-Pyridinecarboxylic acid','5-ethyl-','methyl ester, Picolinic acid','5-ethyl-','methyl ester (7CI,8CI), Methyl 5-ethyl-2-pyridinecarboxylate, 5-Ethyl-2-pyridinecarboxylic acid methyl ester, 5-Ethylpyridine-2-carboxylic acid methyl ester, 5-Ethylpicolinic acid methyl ester, Methyl 5-ethylpicolinate
InChI formel InChI=1S/C9H11NO2/c1-3-7-4-5-8(10-6-7)9(11)12-2/h4-6H,3H2,1-2H3
SMIL CCc1ccc(nc1)C(=O)OC
IUPAC navn methyl 5-ethylpyridine-2-carboxylate
Molekylvægt (g/mol) 165.19
Formel vægt 165.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.