Få mere at vide
Methyl 5-Chloro-3-(chlorosulfonyl)thiophene-2-carboxylate, TRC
Artikelnummer. 30483583
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483583 500 mg
30479018 100 mg
30470550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483583 Leverandør Toronto Research Chemicals Leverandørnr. TRCM219133500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 126910-68-7
Molekylær formel C6H4Cl2O4S2
Molekylvægt (g/mol) 275.11
IUPAC navn methyl 5-chloro-3-chlorosulfonylthiophene-2-carboxylate
SMIL COC(=O)c1sc(Cl)cc1S(=O)(=O)Cl
Tekniske data
Kemisk navn eller materiale Methyl 5-Chloro-3-(chlorosulfonyl)thiophene-2-carboxylate
Mængde 500 mg
InChI formel InChI=1S/C6H4Cl2O4S2/c1-12-6(9)5-3(14(8,10)11)2-4(7)13-5/h2H,1H3
Formel vægt 273.8928