missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromosalicylate, 98+%, Thermo Scientific Chemicals

Catalog Number 15401549
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4068-76-2
C8H7BrO3
231.045
MFCD00016460
FJYDBKPPGRZSOZ-UHFFFAOYSA-N
methyl 5-bromosalicylate, benzoic acid, 5-bromo-2-hydroxy-, methyl ester, 5-bromosalicylic acid methyl ester, methyl 5-bromo-2-hydroxybenzenecarboxylate, methyl 2-hydroxy-5-bromobenzoate, 5-bromo-2-hydroxy-benzoic acid methyl ester, methyl5-bromosalicylate, pubchem19740, acmc-1afpq, methyl-5-bromosalicylate
77683
methyl 5-bromo-2-hydroxybenzoate
COC(=O)C1=C(C=CC(=C1)Br)O
This item is not returnable. View return policy
4068-76-2
C8H7BrO3
231.045
MFCD00016460
FJYDBKPPGRZSOZ-UHFFFAOYSA-N
methyl 5-bromosalicylate, benzoic acid, 5-bromo-2-hydroxy-, methyl ester, 5-bromosalicylic acid methyl ester, methyl 5-bromo-2-hydroxybenzenecarboxylate, methyl 2-hydroxy-5-bromobenzoate, 5-bromo-2-hydroxy-benzoic acid methyl ester, methyl5-bromosalicylate, pubchem19740, acmc-1afpq, methyl-5-bromosalicylate
77683
methyl 5-bromo-2-hydroxybenzoate
COC(=O)C1=C(C=CC(=C1)Br)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 61°C to 65°C (literature)
Color Brown to White
Quantity 5 g
Beilstein 3047854
Formula Weight 231.05
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Methyl 5-bromosalicylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'