Få mere at vide
Methyl 5-Bromo-4-methoxypicolinate, TRC
Artikelnummer. 30475097
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475097 1 g
30483514 5 g
30481418 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475097 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2968031G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1256789-95-3
Molekylær formel C8H8NO3Br
Molekylvægt (g/mol) 246.05
IUPAC navn methyl 5-bromo-4-methoxypyridine-2-carboxylate
SMIL COC(=O)c1cc(OC)c(Br)cn1
Tekniske data
Kemisk navn eller materiale Methyl 5-Bromo-4-methoxypicolinate
Mængde 1 g
InChI formel InChI=1S/C8H8BrNO3/c1-12-7-3-6(8(11)13-2)10-4-5(7)9/h3-4H,1-2H3
Formel vægt 244.9688