Få mere at vide
Methyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate (Gemfibrozil Methyl Ester), Mikromol™
Beskrivelse
Methyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate (Gemfibrozil Methyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate |
| CAS | 149105-25-9 |
| Analyt- eller komponentnavne | Methyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H24 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Methyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate (Gemfibrozil Methyl Ester), Gemfibrozil Imp. I (EP), 5-(2,5-Dimethylphenoxy)-2,2-dimethyl-pentanoic acid methyl ester, Methyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate, Gemfibrozil Impurity I |
| InChI formel | InChI=1S/C16H24O3/c1-12-7-8-13(2)14(11-12)19-10-6-9-16(3,4)15(17)18-5/h7-8,11H,6,9-10H2,1-5H3 |
| SMIL | COC(=O)C(C)(C)CCCOc1cc(C)ccc1C |
| IUPAC navn | methyl 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoate |
| Vis mere |