Få mere at vide
Methyl 4-Methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzoate, TRC
Artikelnummer. 30476013
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476013 100 mg
30470918 1 g
30467296 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476013 Leverandør Toronto Research Chemicals Leverandørnr. TRCM325060100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 917392-54-2
Molekylær formel C18H16N4O2
Molekylvægt (g/mol) 320.35
Synonym Benzoic acid, 4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-, methyl ester, Methyl 4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzoate, Methyl 4-Methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzoate, Nilotinib methyl ester, Methyl 4-Methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]benzoic Acid Ester, Benzoic acid, 4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-, methyl ester, Methyl 4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzoate, Methyl 4-Methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzoate, Nilotinib methyl ester
IUPAC navn methyl 4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]benzoate
SMIL COC(=O)c1ccc(C)c(Nc2nccc(n2)c3cccnc3)c1
Tekniske data
Kemisk navn eller materiale Methyl 4-Methyl-3-[(4-(pyridin-3-yl)pyrimidin-2-yl)amino]benzoate
Mængde 100 mg
InChI formel InChI=1S/C18H16N4O2/c1-12-5-6-13(17(23)24-2)10-16(12)22-18-20-9-7-15(21-18)14-4-3-8-19-11-14/h3-11H,1-2H3,(H,20,21,22)
Formel vægt 320.1273