Få mere at vide
Methyl 4-Methyl-3-[[(2RS)-2-[(1-methylethyl)amino]propanoyl]amino]thiophene-2-carboxylate (Isopropylarticaine), Mikromol™
Beskrivelse
Methyl 4-Methyl-3-[[(2RS)-2-[(1-methylethyl)amino]propanoyl]amino]thiophene-2-carboxylate (Isopropylarticaine)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isopropylarticaine |
| CAS | 1796888-45-3 |
| Analyt- eller komponentnavne | Isopropylarticaine |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H20 N2 O3 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2-Thiophenecarboxylic acid, 4-methyl-3-[[2-[(1-methylethyl)amino]-1-oxopropyl]amino]-, methyl ester, Methyl 4-Methyl-3-[[(2RS)-2-[(1-methylethyl)amino]propanoyl]amino]thiophene-2-carboxylate, Articaine Hydrochloride Imp. E (EP), Isopropylarticaine, Articaine Related Compound E, Articaine Hydrochloride Impurity E, Articaine Impurity E |
| InChI formel | InChI=1S/C13H20N2O3S/c1-7(2)14-9(4)12(16)15-10-8(3)6-19-11(10)13(17)18-5/h6-7,9,14H,1-5H3,(H,15,16) |
| SMIL | COC(=O)c1scc(C)c1NC(=O)C(C)NC(C)C |
| IUPAC navn | methyl 4-methyl-3-[2-(propan-2-ylamino)propanoylamino]thiophene-2-carboxylate |
| Vis mere |