missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 4-Hydroxy Cinnamate, TRC

Artikelnummer. 30465988
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465988 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30465988 Leverandør Toronto Research Chemicals Leverandørnr. TRCM312595500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl 4-Hydroxy Cinnamate
CAS 19367-38-5
Molekylær formel C10 H10 O3
Mængde 500 mg
Synonym 2-Propenoic acid','3-(4-hydroxyphenyl)-','methyl ester','(2E)-, 2-Propenoic acid','3-(4-hydroxyphenyl)-','methyl ester','(E)-, Cinnamic acid','p-hydroxy-','methyl ester','(E)- (8CI), (E)-3-(4-Hydroxyphenyl)acrylic acid methyl ester, (E)-4-Hydroxycinnamic acid methyl ester, (E)-Methyl 3-(4-hydroxyphenyl)acrylate, (E)-Methyl p-hydroxycinnamate, Methyl (2E)-3-(4-hydroxyphenyl)prop-2-enoate, Methyl (2E)-3-(4-hydroxyphenyl)prop-2-enoate, Methyl (E)-3-(4-hydroxyphenyl)-2-propenoate, Methyl (E)-3-(4-hydroxyphenyl)prop-2-enoate, Methyl (E)-4-hydroxycinnamate, Methyl (E)-p-coumarate, Methyl (E)-p-hydroxy-cinnamate, Methyl (E)-p-hydroxycinnamate, Methyl p-hydroxy-(E)cinnamate, Methyl p-hydroxy-trans-cinnamate, Methyl trans-4-hydroxycinnamate, Methyl trans-p-coumarate, trans-Methyl p-hydroxycinnamate, trans-p-Coumaric acid methyl ester, trans-p-Hydroxycinnamic acid methyl ester
InChI formel InChI=1S/C10H10O3/c1-13-10(12)7-4-8-2-5-9(11)6-3-8/h2-7,11H,1H3/b7-4+
SMIL COC(=O)\C=C\c1ccc(O)cc1
IUPAC navn methyl (E)-3-(4-hydroxyphenyl)prop-2-enoate
Molekylvægt (g/mol) 178.18
Formel vægt 178.063
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.