missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Mikromol™

Artikelnummer. 17961946
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17961946 25 mg
17922386 100 mg
30461993 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17961946 Leverandør Mikromol Leverandørnr. MM0084.040025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
CAS 35511-14-9
Analyt- eller komponentnavne Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
Forsendelsestilstand Room Temperature
Molekylær formel C10 H9 N O5 S
Certificeringer/compliance ISO 17025
Synonym Methyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Piroxicam Imp. G (EP), Piroxicam Impurity G
InChI formel InChI=1S/C10H9NO5S/c1-16-10(13)8-9(12)6-4-2-3-5-7(6)17(14,15)11-8/h2-5,11-12H,1H3
SMIL COC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1
IUPAC navn methyl 4-hydroxy-1,1-dioxo-2H-1λ^{6},2-benzothiazine-3-carboxylate
Molekylvægt (g/mol) 255.25
Mængde 25 mg
Formel vægt 255.0201 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.