Få mere at vide
Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide, Mikromol™
Artikelnummer. 30049910
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049910 100 mg
30050852 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049910 Leverandør Mikromol Leverandørnr. MM0247.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide
CAS 59804-25-0
Analyt- eller komponentnavne Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide
Forsendelsestilstand Room Temperature
Molekylær formel C9 H9 N O5 S2
Certificeringer/compliance ISO 17025
Synonym Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide, Tenoxicam Imp. B (EP), Tenoxicam Impurity B
InChI formel InChI=1S/C9H9NO5S2/c1-10-6(9(12)15-2)7(11)8-5(3-4-16-8)17(10,13)14/h3-4,11H,1-2H3
SMIL COC(=O)C1=C(O)c2sccc2S(=O)(=O)N1C
IUPAC navn methyl 4-hydroxy-2-methyl-1,1-dioxothieno[2,3-e]thiazine-3-carboxylate
Molekylvægt (g/mol) 275.3
Mængde 100 mg
Formel vægt 274.9922 g/mol