Få mere at vide
Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Mikromol™
Artikelnummer. 17952746
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17952746 100 mg
17932916 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17952746 Leverandør Mikromol Leverandørnr. MM0084.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
CAS 35511-15-0
Analyt- eller komponentnavne Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
Forsendelsestilstand Room Temperature
Molekylær formel C11 H11 N O5 S
Certificeringer/compliance ISO 17025
Synonym Meloxicam Imp. E (EP), Piroxicam Imp. J (EP), Methyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Meloxicam Impurity E, Piroxicam Impurity J
InChI formel InChI=1S/C11H11NO5S/c1-12-9(11(14)17-2)10(13)7-5-3-4-6-8(7)18(12,15)16/h3-6,13H,1-2H3
SMIL COC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1C
IUPAC navn methyl 4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxylate
Molekylvægt (g/mol) 269.27
Mængde 100 mg
Formel vægt 269.0358 g/mol