Få mere at vide
Methyl 4-Formyl-3-methoxybenzoate, TRC
Artikelnummer. 30474199
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474199 250 mg
30472067 500 mg
30466131 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30474199 Leverandør Toronto Research Chemicals Leverandørnr. TRCM305883250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 74733-24-7
Molekylær formel C10H10O4
Molekylvægt (g/mol) 194.18
Synonym Methyl Ester 4-Formyl-3-methoxybenzoic Acid
IUPAC navn methyl 4-formyl-3-methoxybenzoate
SMIL COC(=O)c1ccc(C=O)c(OC)c1
Tekniske data
Kemisk navn eller materiale Methyl 4-Formyl-3-methoxybenzoate
Mængde 250 mg
InChI formel InChI=1S/C10H10O4/c1-13-9-5-7(10(12)14-2)3-4-8(9)6-11/h3-6H,1-2H3
Formel vægt 194.0579