missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 4-ethoxybenzoate, 98%, Thermo Scientific™

Artikelnummer. 11449116
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11449116 100 g 100g
11429116 5 g 5g
11439116 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11449116 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A17980.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 4-ethoxybenzoate is used in the preparation of 4- ethoxybenzoic acid by reacting with aluminum tribromide. It is involved in the preparation of apricoxib, which is a selective cyclooxygenase inhibitor.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 23676-08-6
Molekylær formel C10H12O3
Molekylvægt (g/mol) 180.20
MDL nummer MFCD00017264
InChI nøgle RNHXTCZZACTEMK-UHFFFAOYSA-N
Synonym 4-ethoxybenzoic acid methyl ester, methyl p-ethoxybenzoate, benzoic acid, 4-ethoxy-, methyl ester, methyl-p-ethoxy benzoate, acmc-20akgg, rarechem al bf 0088, timtec-bb sbb041033, 4-ethoxybenzoic acid, methyl ester
PubChem CID 90231
IUPAC navn methyl 4-ethoxybenzoate
SMIL CCOC1=CC=C(C=C1)C(=O)OC

Tekniske data

Kemisk navn eller materiale Methyl 4-ethoxybenzoate
Smeltepunkt 35°C to 37°C
Kogepunkt 160°C (20 mmHg)
Flammepunkt >110°C (230°F)
Mængde 100 g
Beilstein 2691838
Opløselighedsinformation Soluble in alcohol and ether. Insoluble in water.
Formel vægt 180.21
Procent renhed 98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.