Få mere at vide
Methyl 4-(Dimethylamino)benzoate-D4, TRC
Artikelnummer. 30473215
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30473215 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473215 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30370450MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 4-(Dimethylamino)benzoate-D4
CAS 1175002-06-8
Molekylær formel C10H9D4NO2
Mængde 50 mg
Synonym 4-(Dimethylamino)benzoic-2,3,5,6-d4 Acid Methyl Ester
InChI formel InChI=1S/C10H13NO2/c1-11(2)9-6-4-8(5-7-9)10(12)13-3/h4-7H,1-3H3/i4D,5D,6D,7D
SMIL [2H]c1c([2H])c(C(=O)OC)c([2H])c([2H])c1N(C)C
IUPAC navn methyl 2,3,5,6-tetradeuterio-4-(dimethylamino)benzoate
Molekylvægt (g/mol) 183.24
Formel vægt 183.1197