Learn More
Methyl 4-chlorobutyrate, 98+%
CAS: 3153-37-5 | C5H9ClO2 | 136.58 g/mol
193.50 DKK - 368.39 DKK
Chemical Identifiers
CAS | 3153-37-5 |
---|---|
Molecular Formula | C5H9ClO2 |
Molecular Weight (g/mol) | 136.58 |
MDL Number | MFCD00001003 |
InChI Key | ZZUYIRISBMWFMV-UHFFFAOYSA-N |
Synonym | methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate |
PubChem CID | 76612 |
IUPAC Name | methyl 4-chlorobutanoate |
SMILES | COC(=O)CCCCl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3153-37-5 | |
136.58 | |
ZZUYIRISBMWFMV-UHFFFAOYSA-N | |
76612 | |
COC(=O)CCCCl |
C5H9ClO2 | |
MFCD00001003 | |
methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate | |
methyl 4-chlorobutanoate |
Specifications
3153-37-5 | |
1.1200g/mL | |
59°C | |
98% min. (GC) | |
Cl(CH2)3CO2CH3 | |
MFCD00001003 | |
1.12 | |
Solubility in water: .. Other solubilities: soluble in organic solvents | |
COC(=O)CCCCl | |
136.58 | |
136.58 | |
Liquid |
Colorless | |
175.0°C to 176.0°C | |
Authentic | |
C5H9ClO2 | |
1.4311 to 1.4331 | |
02,278 | |
methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate | |
ZZUYIRISBMWFMV-UHFFFAOYSA-N | |
methyl 4-chlorobutanoate | |
76612 | |
98+% | |
Methyl 4-chlorobutyrate, 98+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with wat
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 221-592-9
missing translation for 'tsca' : TSCA
- Methyl 4-chlorobutyrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.