missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-chloro-3-nitrobenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 78°C to 82°C |
Quantity | 5 g |
Beilstein | 1643729 |
Solubility Information | Soluble in water. |
Formula Weight | 215.59 |
Percent Purity | 98% |
Chemical Name or Material | Methyl 4-chloro-3-nitrobenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'