Få mere at vide
Methyl 4-Butyrylamino-3-methyl-5-nitrobenzoate, Mikromol™
Artikelnummer. 30461821
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461821 4 x 25 mg
30050582 100 mg
30051125 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30461821 Leverandør Mikromol Leverandørnr. MM0939.1200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-Butyrylamino-3-methyl-5-nitrobenzoate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 4-Butyrylamino-3-methyl-5-nitrobenzoate
CAS 152628-01-8
Analyt- eller komponentnavne Methyl 4-Butyrylamino-3-methyl-5-nitrobenzoate
Forsendelsestilstand Room Temperature
Molekylær formel C13 H16 N2 O5
Certificeringer/compliance ISO 17025
Synonym 4-Butyrylamino-3-methyl-5-nitrobenzoic acid methyl ester, Methyl 4-Butyrylamino-3-methyl-5-nitrobenzoate
InChI formel InChI=1S/C13H16N2O5/c1-4-5-11(16)14-12-8(2)6-9(13(17)20-3)7-10(12)15(18)19/h6-7H,4-5H2,1-3H3,(H,14,16)
Løsningstype Neat
SMIL CCCC(=O)Nc1c(C)cc(cc1[N+](=O)[O-])C(=O)OC
IUPAC navn methyl 4-(butanoylamino)-3-methyl-5-nitrobenzoate
Molekylvægt (g/mol) 280.28
Mængde 4 x 25 mg
Formel vægt 280.1059 g/mol