Få mere at vide
Methyl 4-(Butylamino)benzoate, TRC
Artikelnummer. 30240489
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240489 50 mg
30240553 250 mg
30240942 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240489 Leverandør Toronto Research Chemicals Leverandørnr. TRCM27516050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 71839-12-8
Molekylær formel C12 H17 N O2
Molekylvægt (g/mol) 207.27
Synonym Methyl 4-(Butylamino)benzoate, Tetracaine Hydrochloride Imp. C (EP)
IUPAC navn methyl 4-(butylamino)benzoate
SMIL CCCCNc1ccc(cc1)C(=O)OC
Tekniske data
Kemisk navn eller materiale Methyl 4-(Butylamino)benzoate
Mængde 50 mg
InChI formel InChI=1S/C12H17NO2/c1-3-4-9-13-11-7-5-10(6-8-11)12(14)15-2/h5-8,13H,3-4,9H2,1-2H3
Formel vægt 207.1259