Få mere at vide
Methyl 4-(Butylamino)benzoate, Mikromol™
Beskrivelse
Methyl 4-(Butylamino)benzoate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl 4-(Butylamino)benzoate |
| CAS | 71839-12-8 |
| Analyt- eller komponentnavne | Methyl 4-(Butylamino)benzoate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H17 N O2 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Tetracaine Hydrochloride Imp. C (EP), Tetracaine Imp. C (EP), Tetracaine USP Related Compound C, Tetracaine USP RC C, Methyl 4-(Butylamino)benzoate, Tetracaine Related Compound C, Tetracaine Hydrochloride Impurity C, Tetracaine Impurity C |
| InChI formel | InChI=1S/C12H17NO2/c1-3-4-9-13-11-7-5-10(6-8-11)12(14)15-2/h5-8,13H,3-4,9H2,1-2H3 |
| SMIL | CCCCNc1ccc(cc1)C(=O)OC |
| IUPAC navn | methyl 4-(butylamino)benzoate |
| Vis mere |