Få mere at vide
Methyl 4'-(Bromomethyl)biphenyl-2-carboxylate, Mikromol™
Artikelnummer. 17954186
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17954186 100 mg
17973916 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17954186 Leverandør Mikromol Leverandørnr. MM0939.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4'-(Bromomethyl)biphenyl-2-carboxylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 4'-(Bromomethyl)biphenyl-2-carboxylate
CAS 114772-38-2
Analyt- eller komponentnavne Methyl 4'-(Bromomethyl)biphenyl-2-carboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C15 H13 Br O2
Certificeringer/compliance ISO 9001
Synonym Methyl 4'-(Bromomethyl)biphenyl-2-carboxylate
InChI formel InChI=1S/C15H13BrO2/c1-18-15(17)14-5-3-2-4-13(14)12-8-6-11(10-16)7-9-12/h2-9H,10H2,1H3
SMIL COC(=O)c1ccccc1c2ccc(CBr)cc2
IUPAC navn methyl 2-[4-(bromomethyl)phenyl]benzoate
Molekylvægt (g/mol) 305.17
Mængde 100 mg
Formel vægt 304.0099 g/mol