Få mere at vide
Methyl 4-Amino-3,6-dichloropyridine-2-carboxylate, TRC
Artikelnummer. 30470126
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470126 5 mg
30469263 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30470126 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3506005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 4-Amino-3,6-dichloropyridine-2-carboxylate
CAS 350601-39-7
Molekylær formel C7H6Cl2N2O2
Mængde 5 mg
Synonym Methyl 4-amino-3,6-dichloropyridine-2-carboxylate, 2-Pyridinecarboxylic acid, 4-amino-3,6-dichloro-, methyl ester, Methyl 4-amino-3,6-dichloropyridine-2-carboxylate, 2-Pyridinecarboxylic acid, 4-amino-3,6-dichloro-, methyl ester
InChI formel InChI=1S/C7H6Cl2N2O2/c1-13-7(12)6-5(9)3(10)2-4(8)11-6/h2H,1H3,(H2,10,11)
SMIL COC(=O)c1nc(Cl)cc(N)c1Cl
IUPAC navn methyl 4-amino-3,6-dichloropyridine-2-carboxylate
Molekylvægt (g/mol) 221.04
Formel vægt 219.9806