Få mere at vide
Methyl 4-Acetamido-5-chloro-2-ethoxybenzoate, TRC
Artikelnummer. 30483757
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483757 250 mg
30474580 100 mg
30476152 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483757 Leverandør Toronto Research Chemicals Leverandørnr. TRCM293263250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4235-43-2
Molekylær formel C12H14ClNO4
Molekylvægt (g/mol) 271.7
Synonym Methyl 4-(Acetylamino)-5-chloro-2-ethoxybenzoate
IUPAC navn methyl 4-acetamido-5-chloro-2-ethoxybenzoate
SMIL CCOc1cc(NC(=O)C)c(Cl)cc1C(=O)OC
Tekniske data
Kemisk navn eller materiale Methyl 4-Acetamido-5-chloro-2-ethoxybenzoate
Mængde 250 mg
InChI formel InChI=1S/C12H14ClNO4/c1-4-18-11-6-10(14-7(2)15)9(13)5-8(11)12(16)17-3/h5-6H,4H2,1-3H3,(H,14,15)
Formel vægt 271.0611