missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 4-(4-methoxyphenyl)-4-oxobutyrate, TRC

Artikelnummer. 30477092
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30477092 10 mg
30472562 100 mg
30475281 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30477092 Leverandør Toronto Research Chemicals Leverandørnr. TRCM77977810MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 5447-74-5
Molekylær formel C12H14O4
Molekylvægt (g/mol) 222.24
IUPAC navn methyl 4-(4-methoxyphenyl)-4-oxobutanoate
SMIL COC(=O)CCC(=O)c1ccc(OC)cc1

Tekniske data

Kemisk navn eller materiale Methyl 4-(4-methoxyphenyl)-4-oxobutyrate
Mængde 10 mg
InChI formel InChI=1S/C12H14O4/c1-15-10-5-3-9(4-6-10)11(13)7-8-12(14)16-2/h3-6H,7-8H2,1-2H3
Formel vægt 222.0892
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.