missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate, 97%, Thermo Scientific™

Catalog Number p-7032756
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1421261-37-1
C12H12N2O3
232.239
MFCD18384922
ZEDYJJCOGRSZFM-UHFFFAOYSA-N
methyl 4-3-ethyl-1,2,4-oxadiazol-5-yl benzoate, 4-3-ethyl-1,2,4-oxadiazol-5-yl benzic acid methyl ester
73995175
methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate
CCC1=NOC(=N1)C2=CC=C(C=C2)C(=O)OC
This item is not returnable. View return policy
1421261-37-1
C12H12N2O3
232.239
MFCD18384922
ZEDYJJCOGRSZFM-UHFFFAOYSA-N
methyl 4-3-ethyl-1,2,4-oxadiazol-5-yl benzoate, 4-3-ethyl-1,2,4-oxadiazol-5-yl benzic acid methyl ester
73995175
methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate
CCC1=NOC(=N1)C2=CC=C(C=C2)C(=O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 86°C to 91°C
Quantity 250 mg
Formula Weight 232.24
Percent Purity 97%
Chemical Name or Material Methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'